Learn More
Invitrogen™ PAH Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA555840
Description
Immunogen sequence: MSTAVLENPG LGRKLSDFGQ ETSYIEDNCN QNGAISLIFS LKEEVGALAK VLRLFEENDV NLTHIESRPS RLKKDEYEFF THLDKRSLPA LTNIIKILRH DIGATVHELS RDKKKDTVPW FPRTIQELDR FANQILSYGA ELDADHPG Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 88%.
PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
Specifications
| PAH | |
| Polyclonal | |
| Unconjugated | |
| PAH | |
| AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5053 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P00439 | |
| PAH | |
| Recombinant protein corresponding to Human PAH. Recombinant protein control fragment (Product #RP-99363). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.