Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAH Recombinant Rabbit Monoclonal Antibody (JE64-47)
GREENER_CHOICE

Catalog No. FEMA550671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550671 Supplier Invitrogen™ Supplier No. MA550671
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: Rat liver tissue lysates, rat kidney tissue, mouse kidney tissue, HepG2. Predicted band size: 52 kDa Subcellular Location: Cytosol.

PAH encodes the enzyme phenylalanine hydroxylase that is the rate-limiting step in phenylalanine catabolism. Deficiency of this enzyme activity results in the autosomal recessive disorder phenylketonuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAH
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone JE64-47
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene PAH
Gene Accession No. P00439, P04176, P16331
Gene Alias AW106920; Pah; PH; phe-4-monooxygenase; phenylalanine 4-monooxygenase; phenylalanine hydroxylase; phenylalanine-4-hydroxylase; PKU; PKU1
Gene Symbols PAH
Host Species Rabbit
Immunogen Synthetic peptide within human PAH aa 50-100/452.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18478, 24616, 5053
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.