Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAI-2/SerpinB2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS1864R

Description
PAI2 inhibits urokinase-type plasminogen activator. The monocyte derived PAI-2 is distinct from the endothelial cell-derived PAI-1.Specifications
PAI-2/SerpinB2 | |
Polyclonal | |
Unconjugated | |
SERPINB2 | |
HsT1201; Monocyte Arg-serpin; ovalbumin; PAI; Pai2; PAI-2; Pai2a; Placental plasminogen activator inhibitor; PLANH2; Plasminogen activator inhibitor 2; Plasminogen activator inhibitor 2 type A; Plasminogen activator inhibitor 2, macrophage; plasminogen activator inhibitor, type II; plasminogen activator inhibitor, type II (arginine-serpin); serine (or cysteine) peptidase inhibitor, clade B, member 2; serine (or cysteine) proteinase inhibitor, clade B (ovalbumin), member 2; serine (or cysteine) proteinase inhibitor, clade B, member 2; Serpin B2; serpin family B member 2; serpin peptidase inhibitor, clade B (ovalbumin), member 2; SERPINB2; Urokinase inhibitor | |
Rabbit | |
Protein A | |
RUO | |
5055 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P05120 | |
SERPINB2 | |
KLH conjugated synthetic peptide derived from human PAI-2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction