Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAICS Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA550472

Catalog No. PA550472


Only null left
Add to Cart

Description

Description

The antibody detects endogenous levels of total PAICS protein.

This gene encodes a bifunctional enzyme containing phosphoribosylaminoimidazole carboxylase activity in its N-terminal region and phosphoribosylaminoimidazole succinocarboxamide synthetase in its C-terminal region. It catalyzes steps 6 and 7 of purine biosynthesis. The gene is closely linked and divergently transcribed with a locus that encodes an enzyme in the same pathway, and transcription of the two genes is coordinately regulated. The human genome contains several pseudogenes of this gene. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

PAICS
Polyclonal
Unconjugated
PAICS
2610511I09Rik; ADE2; ADE2H1; AIR carboxylase; Airc; Multifunctional protein ADE2; multifunctional protein ADE2H1; Paics; PAIS; Phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase; phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoribosylaminoimidazole succinocarboxamide synthetase; phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoribosylaminoimidazole, succinocarboxamide synthetase; phosphoribosylaminoimidazole carboxylase; phosphoribosylaminoimidazolesuccinocarboxamide synthase; Phosphoribosylaminoimidazole-succinocarboxamide synthase; SAICAR synthetase
Rabbit
Antigen Affinity Chromatography
RUO
10606
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.6 mg/mL
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4
P22234
PAICS
Fusion protein corresponding to residues near the N terminal of human phosphoribosylaminoimidazole carboxylase, phosphoribosylaminoimidazole succinocarboxamide synthetase.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.