Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pain Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$207.00 - $499.50
Specifications
Antigen | Pain |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017944
![]() |
Novus Biologicals
NB100987370.025ML |
0.025 mL |
Each for $207.00
|
|
|||||
NB10098737
![]() |
Novus Biologicals
NB10098737 |
0.1 mL |
Each for $499.50
|
|
|||||
Description
Pain Polyclonal specifically detects Pain in Drosophila samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
Pain | |
Western Blot 1:300-1:2000, Immunohistochemistry 1:300-1:2000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:300-1:2000, Immunohistochemistry-Frozen 1:300-1:2000 | |
Polyclonal | |
Antisera | |
RUO | |
Painless protein, Transient protein painless, Transient receptor potential cation channel protein painless | |
IgG | |
Unpurified | |
Pain (Transient receptor potential cation channel protein painless). |
LYOPH | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Drosophila | |
A synthetic peptide from 3rd extracellular loop of drosophila Pain conjugated to an immunogenic carrier protein was used as the immunogen. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title