Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methanol, For pesticide residue analysis, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products

Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
Description
- For gas chromatography (GC)
- Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
- Non-volatile matter and water content are specified for each solvent
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications
Specifications
Density | 0.791g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.36 |
Flash Point | 9°C (48.2°F) |
Ignition Point | 725°F |
Linear Formula | CH3OH |
Vapor Pressure | 410 mmHg (50°C), 97.68 mmHg (20°C) |
UN Number | UN1230 |
Beilstein | 1098229 |
Formula Weight | 32.04g/mol |
Show More |
Safety and Handling
Hazard Statement | P210-P260-P280-P301 + P310-P311 |
RTECSNumber | PC1400000 |
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction