Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA567412
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA567412 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA567412 Supplier Invitrogen™ Supplier No. PA567412
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RVHATSTYREA TPENDPNYFM NEPTFYTSDG VPFTAADPDY QEKYQELLER EDFFPDYEEN GTDLSGAGDP YLDDIDDEMD Highest antigen sequence identity to the following orthologs - mouse 100%, rat 100%.

The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAIP1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAIP1
Gene Accession No. Q9H074
Gene Alias PABC1-interacting protein 1; PABP-interacting protein 1; Paip1; PAIP-1; poly(A) binding protein interacting protein 1; poly(A)-binding protein-interacting protein 1; polyadenylate binding protein-interacting protein 1; polyadenylate-binding protein-interacting protein 1
Gene Symbols PAIP1
Host Species Rabbit
Immunogen Recombinant Human PAIP1. Recombinant protein control fragment (Product #RP-108074).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10605
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.