Learn More
Invitrogen™ PAIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596495
Description
Immunogen sequence: MSDGFDRAPE QTRPLRAPPS SQDKIPQQNS ESAMAKPQVV VAPVLMSKLS VNAPEFYPSG YSSSYTESYE DGCEDYPTLS EYVQDFLNHL TEQPGSFETE IEQFAETLNG CVTTDDALQE LVELIYQQAT SIPNFSYMGA RLCNYLSHHL TISPQSGNFR QLLLQRCRTE YEVKDQAAKG DEVTRKRFHA FVLFLGELYL NLEIKGTNGQ VTRADILQVG LRELLNALFS NPMDDNLICA VKLLKLTGSV LEDAWKEKGK; Positive Samples: HeLa, HepG2, A-549, 293T, BT-474, Mouse liver, Mouse kidney, Mouse brain, Rat brain; Cellular Location: Cytoplasm.
The protein encoded by this gene interacts with poly(A)-binding protein and with the cap-binding complex eIF4A. It is involved in translational initiation and protein biosynthesis. Overexpression of this gene in COS7 cells stimulates translation. Alternative splicing occurs at this locus and three transcript variants encoding three distinct isoforms have been identified.
Specifications
PAIP1 | |
Polyclonal | |
Unconjugated | |
PAIP1 | |
PABC1-interacting protein 1; PABP-interacting protein 1; Paip1; PAIP-1; poly(A) binding protein interacting protein 1; poly(A)-binding protein-interacting protein 1; polyadenylate binding protein-interacting protein 1; polyadenylate-binding protein-interacting protein 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10605, 218693, 365684 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
1.64 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8VE62, Q9H074 | |
PAIP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human PAIP1 (NP_8775901). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.