Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAIP2 Antibody (1C12) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP222555
Description
PAIP2 Monoclonal specifically detects PAIP2 in Human samples. It is validated for Western Blot, ELISA.Specifications
PAIP2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:1000, ELISA 1:100-1:2000 | |
MGC72018, PABC1-interacting protein 2, PABP-interacting protein 2, PAIP-2, PAIP2APoly(A)-binding protein-interacting protein 2, poly(A) binding protein interacting protein 2, polyA-binding protein-interacting protein 2, polyadenylate-binding protein-interacting protein 2 | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
Anti-human PAIP2 mAb, clone 1C12, is derived from hybridization of mouse F0 myeloma cells with spleen cells from BALB/c mice immunized with a recombinant human PAIP2 protein. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2b κ |
Western Blot, ELISA | |
1C12 | |
Unconjugated | |
NP_057564 | |
PAIP2 | |
Recombinant human PAIP2 (1-127aa) purified from E. coli | |
0.1 mL | |
metabolism | |
51247 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction