Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAIP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563347
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563347 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563347 Supplier Invitrogen™ Supplier No. PA563347
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDPSRSSTSP SIINEDVIIN GHSHEDDNPF AEYM Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

Acts as a repressor in the regulation of translation initiation of poly(A)-containing mRNAs. Its inhibitory activity on translation is mediated via its action on PABPC1. Displaces the interaction of PABPC1 with poly(A) RNA and competes with PAIP1 for binding to PABPC1. Its association with PABPC1 results in disruption of the cytoplasmic poly(A) RNP structure organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAIP2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAIP2
Gene Accession No. Q9BPZ3
Gene Alias 2310050K10Rik; AU045972; HSPC218; MGC72018; PABC1-interacting protein 2; Pabp-interacting protein 2; PAIP2; PAIP-2; PAIP2A; poly(A) binding protein interacting protein 2; poly(A)-binding protein-interacting protein 2; polyA-binding protein-interacting protein 2; polyadenylate-binding protein interacting protein 2; polyadenylate-binding protein-interacting protein 2
Gene Symbols PAIP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAIP2. Recombinant protein control fragment (Product #RP-103268).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.