Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PAIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598807
Description
PAIP2 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Acts as a repressor in the regulation of translation initiation of poly(A)-containing mRNAs. Its inhibitory activity on translation is mediated via its action on PABPC1. Displaces the interaction of PABPC1 with poly(A) RNA and competes with PAIP1 for binding to PABPC1. Its association with PABPC1 results in disruption of the cytoplasmic poly(A) RNP structure organization.
Specifications
PAIP2 | |
Polyclonal | |
Unconjugated | |
PAIP2 | |
2310050K10Rik; AU045972; HSPC218; MGC72018; PABC1-interacting protein 2; Pabp-interacting protein 2; PAIP2; PAIP-2; PAIP2A; poly(A) binding protein interacting protein 2; poly(A)-binding protein-interacting protein 2; polyA-binding protein-interacting protein 2; polyadenylate-binding protein interacting protein 2; polyadenylate-binding protein-interacting protein 2 | |
Rabbit | |
Protein G | |
RUO | |
51247, 67869 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
3.5 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q9BPZ3, Q9D6V8 | |
PAIP2 | |
Recombinant Human Polyadenylate-binding protein-interacting protein 2 protein (1-127AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction