Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK1 Antibody (SR2113), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $413.50
Specifications
Antigen | PAK1 |
---|---|
Clone | SR2113 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunohistochemistry 1:100-1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Classification | Monoclonal |
Description
PAK1 Monoclonal antibody specifically detects PAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, ImmunoprecipitationSpecifications
PAK1 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:30, Immunohistochemistry 1:100-1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunoprecipitation 1:40 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
alpha-PAK, EC 2.7.11, EC 2.7.11.1, MGC130000, MGC130001, p21 protein (Cdc42/Rac)-activated kinase 1, p21/Cdc42/Rac1-activated kinase 1 (STE20 homolog, yeast), p21/Cdc42/Rac1-activated kinase 1 (yeast Ste20-related), p21-activated kinase 1, p65-PAK, PAK-1, PAKalpha, serine/threonine-protein kinase PAK 1, STE20 homolog, yeast | |
A synthesized peptide derived from human PAK1 (Uniprot #: Q13153) | |
Primary | |
Store at -20°C. Avoid freeze/thaw cycles. |
SR2113 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, Neuroscience, Phospho Specific, Signal Transduction | |
PBS (pH 7.4), 150 mM NaCl, 50% Glycerol | |
5058 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title