Learn More
Promega PAK1/CDC42 Kinase Enzyme System, 1mg
Catalog No. PRV6457 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6457 1 mg
1 options
Catalog No. PRV6457 Supplier Promega Supplier No. V6457
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human PAK1 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag and combined with CDC42 at a ratio of 1:4 (PAK1:CDC42) in vitro. PAK1 is a member of the p21-activated kinases (PAKs).
  • Easily Screen and Profile PAK1/CDC42 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C