Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK1 interacting protein 1 Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10077292IR
Description
PAK1 interacting protein 1 Polyclonal antibody specifically detects PAK1 interacting protein 1 in Human samples. It is validated for Western BlotSpecifications
PAK1 interacting protein 1 | |
Polyclonal | |
Western Blot | |
bA421M1.5, FLJ20624, hPIP1WD repeat-containing protein 84, MAK11, p21-activated protein kinase-interacting protein 1, PAK1 interacting protein 1, PIP1PAK/PLC-interacting protein 1, RP11-421M1.5, WDR84PAK1-interacting protein 1 | |
The immunogen recognized by this antibody maps to a region between residue 290 and 340 of human PAK/PLC-interacting protein 1 (WD repeat protein 84) using the numbering given in entry NP_060376.2 (GeneID 55003). | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55003 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction