Learn More
Invitrogen™ PAK1IP1 Polyclonal Antibody

Description
Antibody detects endogenous levels of total PAK1IP1.
Specifications
Specifications
| Antigen | PAK1IP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | PAK1IP1 |
| Gene Accession No. | Q9DCE5, Q9NWT1 |
| Gene Alias | 5830431I15Rik; 5930415H02Rik; AA419825; AI314040; AW556169; bA421M1.5; Gdpd1; glycerophosphodiester phosphodiesterase domain containing 1; hPIP1; MAK11; p21-activated protein kinase-interacting protein 1; PAK/PLC-interacting protein 1; PAK1 interacting protein 1; PAK1-interacting protein 1; PAK1IP1; PIP1; putative PAK inhibitor Skb15; RP11-421M1.5; WD repeat-containing protein 84; WDR84 |
| Gene Symbols | PAK1IP1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.