Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PAK3 Antibody (4G8A5) - BSA Free, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP261767

 View more versions of this product

Catalog No. NBP2-61767


Only null left
Add to Cart

Description

Description

PAK3 Monoclonal specifically detects PAK3 in Human samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

PAK3
Monoclonal
1 mg/mL
Western Blot 1:100-1:2000, ELISA 1:100-1:2000
beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3
Mouse
62.3 kDa
0.1 mL
Primary
Human
Purified
Western Blot, ELISA
4G8A5
Unconjugated
PBS with 0.05% Sodium Azide
PAK3
Purified recombinant fragment of human PAK3 (AA: 1-100) expressed in E. Coli.
Protein G purified
Protein Kinase
5063
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.