Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK3 Antibody (4G8F11) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | PAK3 |
---|---|
Clone | 4G8F11 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
Applications | Western Blot, ELISA |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61766
![]() |
Novus Biologicals
NBP261766 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
PAK3 Monoclonal specifically detects PAK3 in Human, Monkey samples. It is validated for Western Blot, ELISA.Specifications
PAK3 | |
1 mg/mL | |
Western Blot, ELISA | |
Unconjugated | |
Mouse | |
Human, Monkey | |
5063 | |
Purified recombinant fragment of human PAK3 (AA: 1-100) expressed in E. Coli. | |
Primary | |
62.3 kDa |
4G8F11 | |
Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
Monoclonal | |
Purified | |
Protein Kinase | |
beta-PAK, bPAK, CDKN1A, EC 2.7.11, EC 2.7.11.1, hPAK3, mental retardation, X-linked 47, MRX30, MRX47, Oligophrenin-3, OPHN3, p21 (CDKN1A)-activated kinase 3, p21 protein (Cdc42/Rac)-activated kinase 3, p21-activated kinase 3, PAK-3, PAK3beta, serine/threonine-protein kinase PAK 3 | |
PAK3 | |
IgG1 | |
Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title