Learn More
Promega PAK3 Kinase Enzyme System, 1mg
Catalog No. PRV6458 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6458 1 mg
1 options
Catalog No. PRV6458 Supplier Promega Supplier No. V6458
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length mouse PAK3 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. PAK3 is a member of the family of ser/thr p21-activating kinases that serve as targets for the small GTP-binding proteins Cdc42 and RAC.
  • Easily Screen and Profile PAK3 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C