Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAK3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584446
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584446 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584446 Supplier Invitrogen™ Supplier No. PA584446
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DGLDNEEKPP APPLRMNSNN RDSSALNHSS KPLPMAPEEK NKKARLRSIF Highest antigen sequence indentity to the following orthologs: Rat - 98%, Mouse - 98%.

PAK proteins are critical effectors that link Rho GTPases to cytoskeleton reorganization and nuclear signaling. PAK proteins, a family of serine/threonine p21-activating kinases, serve as targets for the small GTP binding proteins Cdc42 and RAC and have been implicated in a wide range of biological activities. The alpha, beta and gammaPAK isoforms complex specifically with Rac1 and Cdc42 in their active GTP-bound state, inhibiting their intrinsic GTPase activity leading to their autophosphorylation. There are eight sites of autophosphorylation on gammaPAK, including Ser 19, Ser 141 and Thr 402, and phosphorylation of Ser 141 and Thr 402 is correlated with gammaPAK activation. Once phosphorylated and their affinity for Rac/Cdc42 reduced, the PAK isoforms disassociate from the complex to seek downstream substrates.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAK3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PAK3
Gene Accession No. O75914
Gene Alias 2.7.11.1; adriamycin resistance-associated; ARA; beta-PAK; bPAK; CDC42/RAC effector kinase PAK-B; CDKN1A; hPAK3; MRX30; MRX47; oligophrenin-3; OPHN3; p21 (CDKN1A)-activated kinase 3; p21 (RAC1) activated kinase 3; p21 protein (Cdc42/Rac)-activated kinase 3; p21/Cdc42/Rac1-activated kinase 3 (yeast Ste20-related); p21-activated kinase 3; p65-PAK; PAK beta; Pak3; Pak-3; PAK3beta; Pak65alpha; Pak65beta; Pakb; Serine/threonine-protein kinase PAK 3; Stk4
Gene Symbols PAK3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PAK3. Recombinant protein control fragment (Product #RP-103370).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5063
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.