Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK4+5+6, p Ser474 Antibody (SR2087), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PAK4+5+6 |
---|---|
Clone | SR2087 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200 |
Applications | Western Blot, Flow Cytometry |
Classification | Monoclonal |
Description
PAK4+5+6 Monoclonal antibody specifically detects PAK4+5+6 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow CytometrySpecifications
PAK4+5+6 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4 | |
A synthesized peptide derived from human phospho-PAK4+5+6 [Ser474 + Ser560 + Ser602] (Uniprot #: O96013, Q9P286, Q9NQU5) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2087 | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Rabbit | |
Phospho Specific | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
10298 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title