Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK4+5+6 Rabbit anti-Human, Mouse, Rat, Clone: 4E1H7, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31692020UL
Description
PAK4+5+6 Monoclonal antibody specifically detects PAK4+5+6 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
PAK4+5+6 | |
Monoclonal | |
Unconjugated | |
EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4 | |
A synthetic phosphorylated peptide around S474 of human PAK4 (O96013). | |
20 μg | |
Phospho Specific | |
10298 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot | |
4E1H7 | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction