Learn More
Promega PAK4 Kinase Enzyme System, 10microg
Catalog No. PRV3201 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV3201 10 μg
1 options
Catalog No. PRV3201 Supplier Promega Supplier No. V3201
Recombinant full-length human PAK4 was expressed by baculovirus in Sf9 cells using an N-terminal GST tag. PAK4 is a member of the p21-activated kinases (PAKs), which have been implicated in the regulation of cell morphology, motility and transformation.
  • Easily Screen and Profile PAK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C