Learn More
PAK4 Mouse anti-Human, Clone: OTI3C3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807368

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The p21-activated kinases (PAKs) are serine-threonine kinases that bind to the active forms of Cdc42 and Rac. They are divided into two groups, the first of which include PAK1, 2 and 3, and can be activated by Cdc42/Rac binding. The second group includes PAK4, 5 and 6. These proteins are not activated by Cdc42/Rac binding. PAK4 was initially identified as a novel effector of Cdc42Hs. Co-expression of PAK4 and Cdc42Hs resulted in induction of filopodia and actin polymerization, showing that it is involved in cytoskeletal reorganization. Other experiments have shown PAK4 to be essential for embryonic viability and proper neuronal development. PAK4 has also been implicated in anchorage-independent growth of tumor cells and is required for activation of several cancer prosurvival pathways.Specifications
PAK4 | |
Monoclonal | |
Unconjugated | |
PAK4 | |
p21 (RAC1) activated kinase 4 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10298 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI3C3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O96013 | |
PAK4 | |
Human recombit protein fragment corresponding to amino acids 1-308 of human PAK4 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.