Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PAK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA520233
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA520233 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA520233 Supplier Invitrogen™ Supplier No. PA520233
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is SW480 cell lysate. PA5-20233 can be used with blocking peptide PEP-0352.

PAK4 is a serine/ threonine p21 activating kinase implicated in the reorganization of actin cytoskeleton and in the formation of filopodia. They are divided into two groups, the first of which include PAK1, 2 and 3, and can be activated by Cdc42/Rac binding. The second group includes PAK4, 5 and 6. These proteins are not activated by Cdc42/Rac binding. PAK4 was initially identified as a novel effector of Cdc42Hs. Co-expression of PAK4 and Cdc42Hs resulted in induction of filopodia and actin polymerization, showing that it is involved in cytoskeletal reorganization. Other experiments have shown PAK4 to be essential for embryonic viability and proper neuronal development. PAK4 has also been implicated in anchorage-independent growth of tumor cells and is required for activation of several cancer prosurvival pathways.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PAK4
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Pak4
Gene Accession No. O96013
Gene Alias 5730488L07Rik; AW555722; hypothetical protein LOC431769; im:5916170; Kiaa1142; mKIAA1142; p21 (CDKN1A)-activated kinase 4; p21 (RAC1) activated kinase 4; p21 protein (Cdc42/Rac)-activated kinase 4; p21(CDKN1A)-activated kinase 4; p21-activated kinase 4; Pak4; PAK-4; protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs; serine/threonine-protein kinase PAK 4; wu:fd12b01; zgc:92014
Gene Symbols Pak4
Host Species Rabbit
Immunogen A 13 amino acid peptide from near the center of human PAK4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10298
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.