Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PAK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$243.00 - $522.00
Specifications
Antigen | PAK4 |
---|---|
Applications | Western Blot, KnockDown |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Description
PAK4 Polyclonal specifically detects PAK4 in Human, Mouse, Rat samples. It is validated for Western Blot, Knockout Validated.Specifications
PAK4 | |
Polyclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
EC 2.7.11, EC 2.7.11.1, KIAA1142, p21 protein (Cdc42/Rac)-activated kinase 4, p21(CDKN1A)-activated kinase 4, p21-activated kinase 4, PAK-4, protein kinase related to S. cerevisiae STE20, effector for Cdc42Hs, serine/threonine-protein kinase PAK 4 | |
Recombinant protein of human PAK4. | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, KnockDown | |
Unconjugated | |
Rabbit | |
Apoptosis, Phospho Specific, Protein Kinase | |
PBS with 50% glycerol, pH7.3. | |
10298 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title