Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Palladium(II) nitrate dihydrate
Shop All MilliporeSigma Sigma Organic Chemistry Products
Click to view available options
Quantity:
1 g
5 g
Description
Palladium(II) nitrate dehydrate is used as a reactant for:Doping activated carbon for catalysisCatalytic use of hydroxyapatite (HAP) supported Pd nanoclusters in the hydrolysis of ammonia-borane.It's used as catalyst for:Selective hydrogenation of 1-heptyne and napthaleneIn highly porous coordination polymer MIL-101 support. It may also be used as a reactant for:Preparation of platinum-palladium/carbon alloy nanocatalysts for methanol-tolerant oxygen reduction reaction in fuel cellsSynthesis of Cu-Pd alloy thin films on Ti substrates by co-electrodeposition of Pd and Cu from nitrate-base electrolytic bathsPreparation of palladium catalyst supported on vertically aligned multi-walled carbon nanotubes for microwave-assisted Heck reactions of p-iodonitrobenzene with styrene and Et acrylatePreparation of di-phenyl sulfide-modified Pd/TiO2 catalysts for acetylene hydrogenation
Specifications
Specifications
| CAS | 32916-07-7 |
| Quantity | 5 g |
| Molecular Formula | N2O6Pd -+ 2H2O |
| Linear Formula | Pd(NO3)2 -+ 2H2O |
| MDL Number | MFCD00149819 |
| Synonym | Palladium dinitrate hydrate |
| Molecular Weight (g/mol) | 266.46 |
| Percent Purity | ∼40%Pd basis |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction