Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALB2 Antibody (M41-P3F3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | PALB2 |
---|---|
Clone | M41-P3F3 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
Applications | Western Blot, ELISA, Immunohistochemistry |
Description
PALB2 Monoclonal antibody specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
PALB2 | |
1 mg/mL | |
Western Blot, ELISA, Immunohistochemistry | |
Unconjugated | |
Mouse | |
Breast Cancer, Cancer | |
PBS | |
79728 | |
IgG1 κ | |
Protein A purified |
M41-P3F3 | |
Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
Peptide Sequence - KENQFLMPPEE | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title