Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALB2 Antibody (M41-P3F3), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250232H
Description
PALB2 Monoclonal specifically detects PALB2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.Specifications
PALB2 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
PALB2 | |
Peptide Sequence - KENQFLMPPEE | |
0.1 mL | |
Breast Cancer, Cancer | |
79728 | |
Store at 4C in the dark. | |
IgG1 κ |
Western Blot, ELISA, Immunohistochemistry | |
M41-P3F3 | |
Western Blot, ELISA, Immunohistochemistry | |
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction