Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALB2 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176317MFV610
Description
PALB2 Polyclonal antibody specifically detects PALB2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
PALB2 | |
Polyclonal | |
Western Blot, ELISA | |
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human PALB2. The immunogen is located within amino acids 940 - 990 of PALB2. Amino Acid Squence: SDDESEKQVLLKSGN | |
0.1 mL | |
Breast Cancer, Cancer | |
79728 | |
Store at 4C in the dark. | |
IgG |
Western Blot, ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction