Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PALB2 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB10060439PCP
Description
PALB2 Polyclonal antibody specifically detects PALB2 in Human, Mouse samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
PALB2 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
DKFZp667I166, DKFZp686E1054, FANCNFLJ21816, partner and localizer of BRCA2, PNCA3 | |
Maps to a region between residue 200 and 250 of human partner and localizer of BRCA2 using the numbering given in entry NP_078951.2 (GeneID 79728). | |
0.1 mL | |
Breast Cancer, Cancer | |
79728 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
PerCP | |
PBS | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction