Learn More
PALB2 Mouse anti-Human, Clone: OTI1E7, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF806541

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The breast cancer early onset proteins BRCA1 and BRCA2 are central to the repair of DNA damage by homologous recombination and are strongly associated with inherited breast and ovarian cancer. The protein PALB2 is required for the localization of BRCA2 to sites of DNA damage and like BRCA1 and 2 is a breast cancer susceptibility gene. PALB2 is thought to function through directly binding to BRCA1, which allows the PALB2 protein to organize BRCA2 and the recombinase RAD51 at the site of DNA damage. Analysis of the BRCA1-PALB2-BRCA2-RAD51network may allow predictions to be made of the responsiveness of a particular tumor to therapeutic treatments. At least four isoforms of PALB2 are known to exist.Specifications
PALB2 | |
Monoclonal | |
Unconjugated | |
PALB2 | |
FANCN, PNCA3 | |
Mouse | |
Affinity Chromatography | |
RUO | |
79728 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1E7 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86YC2 | |
PALB2 | |
Human recombit protein fragment corresponding to amino acids 1-287 of human PALB2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.