Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Paliperidone
GREENER_CHOICESDP

Catalog No. 449350 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-935-0 50 mg
44-931-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 44-935-0 Supplier R&D Systems™ Supplier No. 4493/50
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Atypical antipsychotic; 5-HT2A and D2 antagonist

Atypical antipsychotic; metabolite of risperidone (Cat. No. 2865). Exhibits high affinity for 5-HT2A receptors (Ki = 0.4 nM for cloned human 5-HT2A receptors). Also displays nanomolar affinity for human D2 receptors.

Chemical Identifiers

CAS 144598-75-4
Molecular Formula C23H27FN4O3

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 426.48
Purity >98%
Molecular Weight (g/mol) 426.492
Chemical Name or Material Paliperidone
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.