Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladin Antibody (1E6), PE/Atto594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP125959PEATT594
Description
Palladin Monoclonal specifically detects Palladin in Human, Mouse, Rat, Canine, Chicken samples. It is validated for Flow Cytometry, Knockdown Validated.Specifications
Palladin | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
PALLD | |
Endogenous Palladin immunoprecipitated from chicken embryo fibroblasts. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Chicken | |
IgG1 |
Flow Cytometry, KnockDown | |
1000000 | |
Flow Cytometry, KnockDown Validated | |
CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376 | |
Mouse | |
Protein G purified | |
Cellular Markers, Cytoskeleton Markers, Signal Transduction | |
23022.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction