Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladin Antibody (SR1122), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32263950UL
Description
Palladin Monoclonal antibody specifically detects Palladin in Human samples. It is validated for Western BlotSpecifications
Palladin | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1122 | |
Western Blot 1:500-1:2000 | |
CGI151, CGI-151, FLJ22190, FLJ38193, KIAA0992FLJ39139, MYN, myoneurin, palladin, palladin, cytoskeletal associated protein, PNCA1, Sarcoma antigen NY-SAR-77, SIH002FLJ61376 | |
A synthesized peptide derived from human Palladin (Uniprot #: Q8WX93) | |
50 μg | |
Cell Biology, Cellular Markers, Signal Transduction | |
23022 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction