Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium

Catalog No. 503098728 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
10 Ea.
5 Ea.
Catalog No. Quantity
50-309-8728 1 Ea.
50-309-8236 10 Ea.
50-309-7129 5 Ea.
3 options

Catalog No. 50-309-8728

Supplier: Sigma Aldrich GF529307621EA

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium

Specifications

Melting Point 1554°C
Density 12.02 g/cm3 (literature)
Boiling Point 2970°C
Molecular Formula Pd
Linear Formula Pd
Quantity 1 Ea.
MDL Number MFCD00011167
Synonym Palladium; PD004700
Molecular Weight (g/mol) 106.42
Percent Purity 99.99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.