Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium

Catalog No. 503131010 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-313-1010 5 Ea.
50-313-1008 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-313-1010 Supplier Sigma Aldrich Supplier No. GF902034755EA
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium

Specifications

Melting Point 1554°C
Density 12.02 g/cm3 (literature)
Boiling Point 2970°C
Molecular Formula Pd
Linear Formula Pd
MDL Number MFCD00011167
Quantity 5 Ea.
Synonym Palladium; PD000206
Molecular Weight (g/mol) 106.42
Percent Purity ≥99.95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.