Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium, 99.99%, (trace metal basis), granules, Thermo Scientific™

Catalog No. AC437020050
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC437020050 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC437020050 Supplier Thermo Scientific Chemicals Supplier No. 437020050

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

CAS 7440-05-3
Assay Percent Range 100.00%
Molecular Formula Pd
Linear Formula Pd
Quantity 5g
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES [Pd]
IUPAC Name palladium
Molecular Weight (g/mol) 106.42
PubChem CID 23938
ChEBI CHEBI:33363
Formula Weight 106.4
Grade Trace Metal
Packaging Glass bottle
Form Crystalline Granules
Chemical Name or Material Palladium
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.