Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Palladium On Barium Sulfate

Catalog No. 20572910G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Click to view available options
Quantity:
10 g
50 g

Catalog No. 20-572-910G

Supplier: Sigma Aldrich 20572910G

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Palladium on barium sulfate (Pd/BaSO4), also known as Rosenmund catalyst, is used for the decarboxylation of aliphatic esters for further production of biodiesel. It can also be used in hydrogenation reaction. Pd/BaSO4 may be used as a catalyst reservoir for the Stille cross-coupling reaction between iodobenzene and tributylphenyltin.We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Specifications

Linear Formula Pd
MDL Number MFCD03613605
Quantity 10 g
Synonym Pd/BaSO4; Rosenmund catalyst
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.