Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium black, 99.9% (metals basis)

Catalog No. aa0065903
Click to view available options
Quantity:
1 g
5 g

Catalog No. AA0065903

Supplier: Thermo Scientific Chemicals 00065903

Only null left
Add to Cart

Use as a catalyst in hydrogenation reactions. Other uses include the manufacture of paints, adhesives, fibers and coatings. Basic particles of Palladium black have the tendency to cluster into macroscopic aggregates. | CAS: 7440-05-3 | Pd | 106.42 g/mol

It is used as a catalyst in hydrogenation reactions. Further it finds use in the manufacture of paints, adhesives, fibers and coatings.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5-3-7440
Molecular Formula Pd
Molecular Weight (g/mol) 106.42
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
Synonym black, on carbon, paladio, palladium/carbon, element, charcoal, palladium, element, palladex 600, on charcoal, palladium-activated carbon
PubChem CID 23938
ChEBI CHEBI:33363
IUPAC Name palladium
SMILES [Pd]

Specifications

Color Gray to Silver
Melting Point 1555°C
Boiling Point 3067°C
Appearance Powder
Density 12.02g/cm3 at 20°C
Quantity 1 g
Odor Odorless
Form Powder
Assay Percent Range 99.9% (Metals basis)
Solubility Information Insoluble in water; Soluble in aqua regia and fused alkalies
Chemical Name or Material Palladium black
Show More Show Less
Hazard Category H228: Flammable solid.
DOTInformation DOT Class: 4.1, Packaging Group: II
EINECSNumber 231-115-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.