Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium hydroxide, 99%

Catalog No. p-7050802
Click to view available options
Quantity:
0.25 g
1 g

CAS: 12135-22-7 | H2O2Pd | 140.43 g/mol

Acts as a catalyst for arylation reactions. It is employed in hydrogenolysis of benzyl-nitrogen bonds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12135-22-7
Molecular Formula H2O2Pd
Molecular Weight (g/mol) 140.43
MDL Number MFCD00064599
InChI Key NXJCBFBQEVOTOW-UHFFFAOYSA-L
Synonym palladium hydroxide, palladium ii hydroxide, pearlman's catalyst, pearlmans catalyst, dihydroxypalladium, palladiumhydroxide, pearlman catalyst, peariman's catalyst, pearl man's catalyst, pd oh 2 on carbon
PubChem CID 9942167
SMILES [OH-].[OH-].[Pd++]

Specifications

Color Black
Physical Form Powder
Quantity 0.25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
IUPAC Name palladium(2+);dihydroxide
Formula Weight 140.42
Percent Purity 99%
Odor Odorless
Chemical Name or Material Palladium hydroxide
EINECSNumber 235-219-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.