Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Palladium(II) acetate, 47.5% Pd
Click to view available options
Quantity:
1 g
2 g
10 g
50 g
Packaging:
Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 205.0°C |
Quantity | 50 g |
Linear Formula | (CH3CO2)2Pd |
UN Number | 3261 |
Solubility Information | Solubility in water: insoluble. Other solubilities: soluble in acetone, benzene, methylene dichloride,, chloroform, acetonitrile and diethylether, insoluble in alcohols |
IUPAC Name | palladium(2+);diacetate |
Formula Weight | 224.51 |
Packaging | Glass Bottle |
Chemical Name or Material | Palladium(II) Acetate, 47.5% Pd |
Safety and Handling
Hazard Category | GHS Signal Word: Danger |
Hazard Statement | GHS H Statement Causes serious eye damage. May cause long lasting harmful effects to aquatic life. |
Precautionary Statement | GHS P Statement Wear eye protection/face protection. Immediately call a POISON CENTER or doctor/physician. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. |
EINECSNumber | 222-164-4 |
RTECSNumber | AJ1900000 |
TSCA | TSCA |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction