Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) acetylacetonate --force to the web--, 35% Pd

Catalog No. p-178165
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle

CAS: 14024-61-4 | C10H14O4Pd | 304.64 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14024-61-4
Molecular Formula C10H14O4Pd
Molecular Weight (g/mol) 304.64
MDL Number MFCD00000025 MFCD00000025
InChI Key JKDRQYIYVJVOPF-FDGPNNRMSA-L
Synonym palladium diacetonate, acetylacetone palladium ii salt, bis 2,4-pentanedionato palladium ii
PubChem CID 53384484
IUPAC Name (E)-4-oxopent-2-en-2-olate;palladium(2+)
SMILES [Pd++].C\C([O-])=C\C(C)=O.C\C([O-])=C\C(C)=O

Specifications

Melting Point 190.0°C
Color Brown-Red
Physical Form Solution
Quantity 1 g
Linear Formula (CH3COCH=C(CH3)O)2Pd
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in benzene
Formula Weight 304.61
Percent Purity 35% (Pd)
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Palladium(II) acetylacetonate, 35% Pd
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes serious eye damage.
May cause an allergic skin reaction.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doc
EINECSNumber 237-859-8
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.