Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) bromide, 39.8-40.1% Pd, ACROS Organics™

Click to view available options
Quantity:
2g
10g
Packaging:
Glass bottle
13444-94-5
Br2Pd
266.21
MFCD00011170
INIOZDBICVTGEO-UHFFFAOYSA-L
palladium ii bromide,palladium bromide,palladium bromide pdbr2,palladous bromide,palladium ii bromide, premion,palladium 2+ dibromide,pdbr2,palladiumbromide,palladiumbromide pdbr2
83469
dibromopalladium
Br[Pd]Br
13444-94-5
Br2Pd
266.21
MFCD00011170
INIOZDBICVTGEO-UHFFFAOYSA-L
palladium ii bromide,palladium bromide,palladium bromide pdbr2,palladous bromide,palladium ii bromide, premion,palladium 2+ dibromide,pdbr2,palladiumbromide,palladiumbromide pdbr2
83469
dibromopalladium
Br[Pd]Br
Linear Formula PdBr2
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hydrobromic acid
Formula Weight 266.21
Physical Form Powder
Percent Purity 39.8-40.1% Pd
Calcium (Ca) 10ppm max.
Copper (Cu) 10ppm max.
Gold (Au) 20ppm max.
Iridium (Ir) 20ppm max.
Iron (Fe) 10ppm max.
Magnesium (Mg) 10ppm max.
Packaging Glass bottle
Platinum (Pt) 20ppm max.
Rhodium (Rh) 20ppm max.
Ruthenium (Ru) 20ppm max.
Silicon (Si) 50ppm max.
Silver (Ag) 20ppm max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in hydrobromic acid
Color Black to Brown-Red
Quantity 10g
Chemical Name or Material Palladium(II) bromide, 39.8-40.1% Pd
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 236-588-2

TSCA : TSCA