Learn More
Palladium hydroxide, Pd 20% on carbon, nominally 50% water, Pearlman's Catalyst
Description
Hydrogenolysis of benzyl-nitrogen bondsPalladium hydroxide, Pd 20% on carbon, nominally 50% water is used as a catalyst for hydrogenolysis of benzyl groups and epoxides. It is used to reduce nitro compounds to amines. It acts as a dehydrogenation reagent as well as a useful reagent for debenzylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as catalyst for hydrogenolysis of benzyl groups and epoxides.
It is used to reduce nitro compounds to amines.
Acts as dehydrogenation reagent as well as useful reagent for debenzylation

Specifications
Specifications
Color | Black |
Physical Form | Moist Powder |
Quantity | 10 g |
Solubility Information | Insoluble in water. |
Appearance | Black moist powder |
Chemical Name or Material | Palladium hydroxide |
Safety and Handling
Hazard Category | H228 |
Precautionary Statement | P210-P240-P241-P280-P370+P378q |
EINECSNumber | 235-219-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.