Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) chloride, 59% Pd, ACROS Organics™

Click to view available options
Quantity:
1g
5g
100g
Packaging:
Glass bottle
7647-10-1
Cl2Pd
177.31
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
24290
CHEBI:53434
dichloropalladium
Cl[Pd]Cl
7647-10-1
Cl2Pd
177.31
PIBWKRNGBLPSSY-UHFFFAOYSA-L
palladium chloride,palladium ii chloride,palladium dichloride,palladous chloride,pdcl2,palladium 2+ chloride,palladium chloride pdcl2,enplate activator 440
24290
CHEBI:53434
dichloropalladium
Cl[Pd]Cl
Linear Formula PdCl2
Fieser 01,782; 02,303; 04,369; 05,500; 06,447; 07,277; 08,384; 09,352; 10,300; 11,393; 12,371; 13,9; 15,248; 16,268
Merck Index 15,7091
Solubility Information Solubility in water: 31.99g/L. Other solubilities: soluble in hydrochloric acid,hydrobromic acid,soluble in alcohol and acetone
Formula Weight 177.31
Physical Form Powder
Percent Purity 59 to 60% (Pd)
Calcium (Ca) 20ppm max.
Copper (Cu) 15ppm max.
Gold (Au) 30ppm max.
Iridium (Ir) 30ppm max.
Iron (Fe) 15ppm max.
Lead (Pb) 15ppm max.
Magnesium (Mg) 15ppm max.
Packaging Glass bottle
Platinum (Pt) 100ppm max.
Rhodium (Rh) 30ppm max.
Ruthenium (Ru) 30ppm max.
Silicon (Si) 30ppm max.
Silver (Ag) 30ppm max.
Solubility Solubility in water: insoluble. Other solubilities: soluble in hydrochloric acid, hydrobromic acid, soluble in alcohol and acetone
Color Red to Brown
Melting Point 678.0°C to 680.0°C
Quantity 5g
Chemical Name or Material Palladium(II) chloride
Show More Show Less

GHS H Statement
May be corrosive to metals. May cause an allergic skin reaction. Causes serious eye damage.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Absorb spillage to prevent material damage. Keep only in original container.

Danger