Palladium(II) chloride, 99.9% (metals basis), Pd 59.5% min, Alfa Aesar™

For Use As a Catalyst Precursor

Manufacturer: Alfa Aesar 1103418

Quantity 50g

 View more versions of this product

Catalog No. AA1103418

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7647-10-1
    Physical Form Powder
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula PdCl2
    Formula Weight 177.31
    MDL Number MFCD00003558
    Chemical Name or Material Palladium(II) chloride

    Specifications

    Physical Form Powder
    Formula Weight 177.31
    Quantity 50g
    Color Red-brown
    Density 4g/cm3 at 20°C
    EINECS Number 231-596-2
    Odor Odorless
    Assay Percent Range 99.9% (metals basis)
    Molecular Formula PdCl2
    Appearance Red-brown powder
    MDL Number MFCD00003558
    Health Hazard 1 H318-H290-H317-H303: Causes serious eye damage. May be corrosive to metals. May cause an allergic skin reaction. May be harmful if swallowed.
    CAS 7647-10-1
    Chemical Name or Material Palladium(II) chloride

    H318-H290-H317-H303: Causes serious eye damage. May be corrosive to metals. May cause an allergic skin reaction. May be harmful if swallowed.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit