Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) chloride, Premion™, 99.999% (metals basis), Pd 59.5% min

Catalog No. p-7058708
Click to view available options
Quantity:
1 g
5 g

CAS: 7647-10-1 | Cl2Pd | 177.32 g/mol

Palladium(II) chloride is used to remove the stain present stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tatooing of leukomas in thecornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-10-1
Molecular Formula Cl2Pd
Molecular Weight (g/mol) 177.32
MDL Number MFCD00003558
InChI Key PIBWKRNGBLPSSY-UHFFFAOYSA-L
Synonym palladium chloride, palladium ii chloride, palladium dichloride, palladous chloride, pdcl2, palladium 2+ chloride, palladium chloride pdcl2, enplate activator 440
PubChem CID 24290
ChEBI CHEBI:53434
SMILES [Cl-].[Cl-].[Pd++]

Specifications

Melting Point 675°C (decomposition)
Color Red to Brown
Physical Form Powder
Quantity 1 g
Merck Index 14,6990
Solubility Information Soluble in cold water,organic solvents and hydrochloric acid.
IUPAC Name dichloropalladium
Formula Weight 177.31
Percent Purity 99.999%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 59.5% min
Odor Odorless
Density 4.0 g/mL
Assay (metals basis)
Chemical Name or Material Palladium (II) chloride
Show More Show Less
Hazard Category H290-H302-H317-H318
Hazard Statement GHS H Statement
H318-H290-H317
Causes serious eye damage.
May be corrosive to metals.
May cause an allergic skin reaction.
Precautionary Statement P234-P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 231-596-2
RTECSNumber RT3500000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.