Learn More
Palladium(II) chloride, solution, Pd 9.0-11.0% w/w (cont. Pd), Thermo Scientific Chemicals
Description
Palladium(II) chloride is used to remove the stain present stainless steel . Its solutions are used to test the corrosion-resistance of stainless steel. It is used in carbon monoxide detectors. It is also used for cosmetic tatooing of leukomas in thecornea and in the preparation of palladium(II) iodide by reacting with potassium iodide. It is involved in the synthesis of semiconducting metal-containing polymers. It is used as a catalyst for carbonylation of organic tellurides and deamination of phenethylamines to phenyl substituted pyrroles with copper(I) as co catalyst. It plays an important role in the hydrodimerization of butadiene to form octa-2,7-dien-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Brown |
UN Number | UN3264 |
Concentration | Pd 9.0 to 11.0% w/w (cont. Pd) |
Solubility Information | Miscible with water,organic solvents and hydrochloric acid. |
Formula Weight | 177.31 |
Odor | Acidic |
Quantity | 1 g |
Physical Form | Liquid |
Chemical Name or Material | Palladium(II) chloride, Solution |
Safety and Handling
Hazard Category | H290-H317-H318 |
Hazard Statement | GHS H Statement H314-H318-H290-H302-H317 Causes severe skin burns and eye damage. Causes serious eye damage. May be corrosive to metals. Harmful if swallowed. May cause an allergic skin reaction. |
Precautionary Statement | P234-P261-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P363-P390-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S. |
EINECSNumber | 231-596-2 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.