Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium(II) trifluoroacetate, 97%

Catalog No. AA3944806
Click to view available options
Quantity:
1 g
5 g

CAS: 42196-31-6 | C4F6O4Pd | 332.45 g/mol

Palladium(II) trifluoroacetate is used in the preparation of allyl-palladium complexes. It acts as a catalyst used in the mild decarboxylation of electron-rich aromatic acids and the direct cross-coupling of unactivated arenes. Further, it is involved in the selective allylic oxidation of geranylacetone and other olefins to their allyl acetates in order to prepare keto alcohols.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42196-31-6
Molecular Formula C4F6O4Pd
Molecular Weight (g/mol) 332.45
MDL Number MFCD00013204
InChI Key PBDBXAQKXCXZCJ-UHFFFAOYSA-L
Synonym palladium ii trifluoroacetate, palladium ii 2,2,2-trifluoroacetate, palladium 2+ ion ditrifluoroacetate, palladium trifluoroacetate, palladium 2+ ditrifluoroacetate, trifluoroacetic acid palladium ii salt, bis 2,2,2-trifluoroacetoxy palladium, acmc-209jo7, ksc235m3d
PubChem CID 2733347
SMILES [Pd++].[O-]C(=O)C(F)(F)F.[O-]C(=O)C(F)(F)F

Specifications

Melting Point 210°C (decomposition)
Color Brown
Physical Form Powder
Quantity 5 g
Assay Percent Range 97%
Solubility Information Soluble in diethyl ether and acetone. Insoluble in benzene,chloroform and trifluoroacetic acid.
IUPAC Name palladium(2+);2,2,2-trifluoroacetate
Formula Weight 332.44
Sensitivity Hygroscopic
Appearance Brown powder
Chemical Name or Material Palladium(II) trifluoroacetate
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement In case of contact with eyes, rinse immediately with plenty of water and seek medical advice.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.