Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Palladium standard solution, for AAS, 1 mg/ml Pd in 10 to 20% HCl

Catalog No. p-220882
Click to view available options
Quantity:
100 mL
500 mL

CAS: 7440-05-3 | Pd | 106.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For atomic adsorption spectroscopy
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Palladium standard solution
Name Note 1 mg/ml Pd in 10% HCl
Packaging Plastic bottle
CAS 7440-05-3, 7647-01-0, 7732-18-5
Density 1.0500g/mL
Specific Gravity 1.05
Physical Form Solution
Molecular Formula Pd
Concentration 997 to 1003 μg/mL of Pd (2+) in 10% HCl (20°C)
MDL Number MFCD00011167 MFCD03457879 MFCD03427452 MFCD03613602 MFCD03613603 MFCD03613604 MFCD03613605 MFCD06798745
Color Brown/Red
Melting Point -25.0°C
Boiling Point 107.0°C
Solubility Soluble in water
InChI Key KDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES [Pd]
IUPAC Name palladium
Molecular Weight (g/mol) 106.42
Quantity 100 mL
PubChem CID 23938
ChEBI CHEBI:33363
Grade AAS
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.